Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993760.2 | XP_008992008.1 | 1 - 2 | 735 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGBD1| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993760.2 | XP_008992008.1 | 1 - 2 | 735 | 84 | |