Assay Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009007750.2 | XP_009005998.1 | 3 - 4 | 835 | 95 | |
XM_017977937.1 | XP_017833426.1 | 3 - 4 | 835 | 95 | ||
XM_017977938.1 | XP_017833427.1 | 3 - 4 | 839 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
