Assay Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748349.3 | XP_002748395.1 | 4 - 5 | 816 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHRS13| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748349.3 | XP_002748395.1 | 4 - 5 | 816 | 58 | |
Set Membership: |
|