Assay Details
Gene Symbol: NACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743460.4 | XP_002743506.1 | 6 | 5371 | 104 | |
XM_017977373.1 | XP_017832862.1 | 6 | 5246 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
