Assay Details
| Gene Symbol: HPGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745206.4 | XP_002745252.1 | 3 - 4 | 779 | 102 | |
| XM_017970134.1 | XP_017825623.1 | 3 - 4 | 779 | 102 | ||
More Information
Set Membership: |
|

HPGD
Genomic Map