Assay Details
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746741.2 | XP_002746787.1 | 2 - 3 | 703 | 85 | |
XM_008994677.1 | XP_008992925.1 | 2 - 3 | 703 | 85 | ||
XM_008994678.2 | XP_008992926.1 | 2 - 3 | 703 | 85 | ||
XM_017971220.1 | XP_017826709.1 | 2 - 3 | 703 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
