Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008995289.2 | XP_008993537.1 | 5 - 6 | 587 | 68 | |
XM_017971489.1 | XP_017826978.1 | 6 - 7 | 610 | 68 | ||
XM_017971490.1 | XP_017826979.1 | 6 - 7 | 607 | 68 | ||
XM_017971491.1 | XP_017826980.1 | 9 - 10 | 716 | 68 | ||
XM_017971492.1 | XP_017826981.1 | 8 - 9 | 1666 | 68 | ||
XM_017971493.1 | XP_017826982.1 | 8 - 9 | 639 | 68 | ||
XM_017971495.1 | XP_017826984.1 | 10 - 11 | 789 | 68 | ||
XM_017971496.1 | XP_017826985.1 | 9 - 10 | 1739 | 68 | ||
XM_017971497.1 | XP_017826986.1 | 9 - 10 | 712 | 68 | ||
XM_017971498.1 | XP_017826987.1 | 9 - 10 | 714 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
