Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748201.3 | XP_002748247.1 | 4 - 5 | 865 | 73 | |
XM_008997115.2 | XP_008995363.1 | 4 - 5 | 821 | 73 | ||
XM_008997116.2 | XP_008995364.1 | 4 - 5 | 855 | 73 | ||
XM_008997117.2 | XP_008995365.1 | 6 - 7 | 960 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
