Assay Details
Gene Symbol: CARD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762318.3 | XP_002762364.2 | 12 | 1827 | 76 | |
XM_008988319.2 | XP_008986567.1 | 12 | 3212 | 76 | ||
XM_008988320.2 | XP_008986568.1 | 12 | 1862 | 76 | ||
XM_008988321.2 | XP_008986569.1 | 11 | 3090 | 76 | ||
XM_008988322.2 | XP_008986570.1 | 11 | 3059 | 76 | ||
XM_008988323.2 | XP_008986571.1 | 13 | 2035 | 76 | ||
XM_017968105.1 | XP_017823594.1 | 12 | 3210 | 76 | ||
XM_017968107.1 | XP_017823596.1 | 9 | 2614 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
