Assay Details
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205139.2 | NP_001192068.2 | 2 - 3 | 323 | 81 | |
NM_001278383.1 | NP_001265312.1 | 2 - 3 | 326 | 81 | ||
GenBank mRNA | HX460499.1 | - | 2 - 3 | 324 | 81 | |
HX584077.1 | - | 1 - 2 | 76 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
