Assay Details
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205139.2 | NP_001192068.2 | 4 - 5 | 461 | 70 | |
NM_001278383.1 | NP_001265312.1 | 4 - 5 | 464 | 70 | ||
GenBank mRNA | HX460499.1 | - | 4 - 5 | 462 | 70 | |
HX584077.1 | - | 3 - 4 | 214 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
