Assay Details
| Gene Symbol: LOC100389947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756462.4 | XP_002756508.1 | 5 - 6 | 865 | 108 | |
| GenBank mRNA | KJ922561.1 | - | 5 - 6 | 865 | 108 | |
More Information
Set Membership: |
|

LOC100389947
Genomic Map