Assay Details
Gene Symbol: LOC100389947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756462.4 | XP_002756508.1 | 7 - 8 | 1201 | 83 | |
GenBank mRNA | KJ922561.1 | - | 7 - 8 | 1201 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
