Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257229.1 | NP_001244158.1 | 3 - 4 | 439 | 61 | |
| XM_017971638.1 | XP_017827127.1 | 4 - 5 | 961 | 61 | ||
| GenBank mRNA | AF272013.3 | - | 3 - 4 | 439 | 61 | |
More Information
Set Membership: |
|

HSD17B1
Genomic Map