Assay Details
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758919.4 | XP_002758965.1 | 4 - 5 | 1283 | 74 | |
XM_008982674.2 | XP_008980922.1 | 5 - 6 | 1277 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
