Assay Details
| Gene Symbol: CYBRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749356.2 | XP_002749402.1 | 3 - 4 | 860 | 74 | |
| XM_008998720.1 | XP_008996968.1 | 2 - 3 | 651 | 74 | ||
More Information
Set Membership: |
|

CYBRD1
Genomic Map