Assay Details
Gene Symbol: PPCDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753350.4 | XP_002753396.1 | 5 - 6 | 690 | 69 | |
XM_009005275.2 | XP_009003523.1 | 5 - 6 | 760 | 69 | ||
XM_009005276.2 | XP_009003524.1 | 3 - 4 | 465 | 69 | ||
XM_009005277.2 | XP_009003525.1 | 4 - 5 | 452 | 69 | ||
XM_017976763.1 | XP_017832252.1 | 4 - 5 | 698 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
