Assay Details
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017974437.1 | XP_017829926.1 | 4 - 5 | 1109 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
