Assay Details
Gene Symbol: LOC100415620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752728.4 | XP_002752774.2 | 7 - 8 | 784 | 106 | |
XM_009004180.2 | XP_009002428.1 | 7 - 8 | 673 | 106 | ||
XM_009004181.2 | XP_009002429.1 | 7 - 8 | 673 | 106 | ||
XM_017976194.1 | XP_017831683.1 | 7 - 8 | 588 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
