Assay Details
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760629.3 | XP_002760675.2 | 3 - 4 | 299 | 66 | |
XM_008985378.2 | XP_008983626.1 | 4 - 5 | 543 | 66 | ||
XM_008985380.2 | XP_008983628.1 | 4 - 5 | 463 | 66 | ||
XM_008985381.1 | XP_008983629.1 | 3 - 4 | 270 | 66 | ||
XM_008985382.1 | XP_008983630.1 | 3 - 4 | 374 | 66 | ||
XM_008985384.1 | XP_008983632.1 | 3 - 4 | 308 | 66 | ||
XM_008985387.2 | XP_008983635.1 | 3 - 4 | 448 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
