Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008987814.2 | XP_008986062.1 | 3 - 4 | 366 | 79 | |
XM_008987815.2 | XP_008986063.1 | 3 - 4 | 295 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
