Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989812.2 | XP_008988060.1 | 3 - 4 | 695 | 86 | |
| XM_017968597.1 | XP_017824086.1 | 3 - 4 | 695 | 86 | ||
More Information
Set Membership: |
|

IL13RA1
Genomic Map