Assay Details
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756298.4 | XP_002756344.1 | 7 - 8 | 2468 | 61 | |
XM_009009819.2 | XP_009008067.1 | 8 - 9 | 2580 | 61 | ||
XM_017979061.1 | XP_017834550.1 | 8 - 9 | 2522 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
