Assay Details
Gene Symbol: MINDY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750063.3 | XP_002750109.1 | 7 - 8 | 883 | 74 | |
XM_008999982.1 | XP_008998230.1 | 7 - 8 | 883 | 74 | ||
XM_017974078.1 | XP_017829567.1 | 6 - 7 | 837 | 74 | ||
XM_017974079.1 | XP_017829568.1 | 6 - 7 | 782 | 74 | ||
XM_017974080.1 | XP_017829569.1 | 5 - 6 | 730 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
