Assay Details
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984852.2 | XP_008983100.1 | 3 - 4 | 434 | 70 | |
XM_008984853.2 | XP_008983101.1 | 3 - 4 | 434 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
