Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758203.3 | XP_002758249.1 | 2 - 3 | 1018 | 63 | |
| XM_003734814.3 | XP_003734862.1 | 3 - 4 | 360 | 63 | ||
| XM_008981406.2 | XP_008979654.1 | 2 - 3 | 326 | 63 | ||
| XM_017964777.1 | XP_017820266.1 | 2 - 3 | 1023 | 63 | ||
| XM_017964778.1 | XP_017820267.1 | 2 - 3 | 214 | 63 | ||
| XM_017964779.1 | XP_017820268.1 | 3 - 4 | 361 | 63 | ||
More Information
Set Membership: |
|

FGF12
Genomic Map