Assay Details
Gene Symbol: LRFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762031.3 | XP_002762077.1 | 2 - 3 | 2568 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
