Assay Details
Gene Symbol: ZNF268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009004951.2 | XP_009003199.1 | 5 - 6 | 738 | 84 | |
XM_009004953.2 | XP_009003201.1 | 5 - 6 | 678 | 84 | ||
XM_017976531.1 | XP_017832020.1 | 5 - 6 | 628 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
