Assay Details
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759764.4 | XP_002759810.1 | 3 - 4 | 563 | 68 | |
XM_017965988.1 | XP_017821477.1 | 3 - 4 | 469 | 68 |

More Information
Set Membership: |
![]() ![]() |
