Assay Details
| Gene Symbol: TRMT61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757905.3 | XP_002757951.1 | 3 - 4 | 1702 | 118 | |
| XM_008980947.2 | XP_008979195.1 | 3 - 4 | 1725 | 118 | ||
More Information
Set Membership: |
|

TRMT61B
Genomic Map