Assay Details
| Gene Symbol: PKIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994964.2 | XP_008993212.1 | 6 - 7 | 587 | 99 | |
| XM_008994965.2 | XP_008993213.1 | 5 - 6 | 515 | 99 | ||
| XM_008994967.2 | XP_008993215.1 | 5 - 6 | 485 | 99 | ||
| XM_008994968.2 | XP_008993216.1 | 4 - 5 | 413 | 99 | ||
| XM_008994971.1 | XP_008993219.1 | 4 - 5 | 413 | 99 | ||
| XM_017971346.1 | XP_017826835.1 | 5 - 6 | 565 | 99 | ||
More Information
Set Membership: |
|

PKIB
Genomic Map