Assay Details
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761765.3 | XP_002761811.2 | 1 - 2 | 263 | 55 | |
XM_008987310.1 | XP_008985558.1 | 1 - 2 | 230 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
