Assay Details
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759855.4 | XP_002759901.1 | 2 | 465 | 72 | |
XM_008984249.1 | XP_008982497.1 | 2 | 433 | 72 | ||
XM_008984250.2 | XP_008982498.1 | 2 | 425 | 72 | ||
GenBank mRNA | EF214954.1 | - | 1 | 318 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
