Assay Details
| Gene Symbol: C15H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758341.2 | XP_002758387.1 | 1 - 2 | 643 | 110 | |
| XM_017964903.1 | XP_017820392.1 | 2 - 3 | 549 | 110 | ||
More Information
Set Membership: |
|

C15H3orf62
Genomic Map