Assay Details
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744169.3 | XP_002744215.1 | 1 - 2 | 384 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
