Assay Details
Gene Symbol: TSPAN17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744506.3 | XP_002744552.3 | 9 | 2354 | 128 | |
XM_017969643.1 | XP_017825132.1 | 9 | 2254 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
