Assay Details
Gene Symbol: UCMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017974062.1 | XP_017829551.1 | 3 - 4 | 917 | 81 | |
XM_017974063.1 | XP_017829552.1 | 3 - 4 | 917 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
