Assay Details
| Gene Symbol: TYW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981271.2 | XP_008979519.1 | 5 - 6 | 740 | 58 | |
| XM_008981273.2 | XP_008979521.1 | 5 - 6 | 722 | 58 | ||
| XM_008981274.2 | XP_008979522.1 | 5 - 6 | 740 | 58 | ||
| XM_008981282.2 | XP_008979530.1 | 4 - 5 | 566 | 58 | ||
| XM_017965926.1 | XP_017821415.1 | 5 - 6 | 589 | 58 | ||
More Information
Set Membership: |
|

TYW1
Genomic Map