Assay Details
| Gene Symbol: MRPL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754794.4 | XP_002754840.1 | 5 - 6 | 1029 | 77 | |
| XM_017977801.1 | XP_017833290.1 | 5 - 6 | 535 | 77 | ||
More Information
Set Membership: |
|

MRPL48
Genomic Map