Assay Details
Gene Symbol: CAPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009004246.2 | XP_009002494.1 | 14 - 15 | 1416 | 87 | |
XM_009004247.2 | XP_009002495.1 | 14 - 15 | 1460 | 87 | ||
XM_009004249.2 | XP_009002497.1 | 13 - 14 | 1341 | 87 | ||
XM_009004251.2 | XP_009002499.1 | 14 - 15 | 1416 | 87 | ||
XM_017976215.1 | XP_017831704.1 | 14 - 15 | 1317 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
