Assay Details
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762386.4 | XP_002762432.3 | 2 - 3 | 1087 | 85 | |
XM_008988430.2 | XP_008986678.1 | 3 - 4 | 617 | 85 | ||
XM_008988432.2 | XP_008986680.1 | 3 - 4 | 842 | 85 | ||
XM_008988433.2 | XP_008986681.1 | 2 - 3 | 1510 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
