Assay Details
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008990413.2 | XP_008988661.2 | 6 - 7 | 2265 | 67 | |
XM_017969648.1 | XP_017825137.1 | 6 - 7 | 2265 | 67 | ||
XM_017969649.1 | XP_017825138.1 | 6 - 7 | 2265 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
