Assay Details
Gene Symbol: SMIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748738.3 | XP_002748784.1 | 1 - 2 | 2116 | 67 | |
XM_008997761.2 | XP_008996009.1 | 2 - 3 | 449 | 67 | ||
XM_008997762.2 | XP_008996010.1 | 2 - 3 | 1423 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
