Assay Details
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756428.4 | XP_002756474.2 | 4 - 5 | 688 | 72 | |
XM_009010240.2 | XP_009008488.1 | 4 - 5 | 688 | 72 | ||
XM_017979290.1 | XP_017834779.1 | 4 - 5 | 688 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
