Assay Details
Gene Symbol: FAM228B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008981067.1 | XP_008979315.1 | 8 - 9 | 1475 | 59 | |
XM_008981072.1 | XP_008979320.1 | 7 - 8 | 1367 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
