Assay Details
Gene Symbol: SMIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009009110.2 | XP_009007358.1 | 3 - 4 | 133 | ||
XM_009009111.2 | XP_009007359.1 | 3 - 4 | 326 | 118 | ||
XM_017978706.1 | XP_017834195.1 | 2 - 3 | 133 | |||
XM_017978707.1 | XP_017834196.1 | 2 - 3 | 284 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
