Assay Details
Gene Symbol: IL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267771.1 | NP_001254700.1 | 2 - 3 | 235 | 66 | |
XM_017975106.1 | XP_017830595.1 | 3 - 4 | 663 | 66 | ||
GenBank mRNA | DQ658153.1 | - | 2 - 3 | 235 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
