Assay Details
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758212.4 | XP_002758258.1 | 4 - 5 | 986 | 72 | |
XM_008981355.2 | XP_008979603.1 | 4 - 5 | 1174 | 72 | ||
XM_017964743.1 | XP_017820232.1 | 3 - 4 | 830 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
