Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008983389.2 | XP_008981637.1 | 2 - 3 | 1297 | 119 | |
XM_017965644.1 | XP_017821133.1 | 3 - 4 | 298 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
