Assay Details
Gene Symbol: LOC100394788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008991860.2 | XP_008990108.1 | 14 - 15 | 2571 | 91 | |
XM_008991864.2 | XP_008990112.1 | 14 - 15 | 2571 | 91 | ||
XM_008991868.2 | XP_008990116.1 | 14 - 15 | 2592 | 91 | ||
XM_008991872.2 | XP_008990120.1 | 12 - 13 | 2497 | 91 | ||
XM_017971364.1 | XP_017826853.1 | 11 - 12 | 1966 | 91 | ||
XM_017971370.1 | XP_017826859.1 | 10 - 11 | 1874 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
